5-Bromothiazole 97%

Supplier: Ambeed
Danger

A145518-100G A145518-10G A145518-250MG A145518-1G A145518-5G
103803-548EA 11 USD
103803-548 103841-632 76997-030 76997-032 76997-034 77299-458 77299-460
5-Bromothiazole 97%
5-Bromothiazole

Precaución: for Research Use Only

Formula: C₃H₂BrNS
Boiling Pt: 197.4 °C (760 mm Hg)
Flash Pt: 76.7 °C
Storage Temperature: Ambient
MDL Number: MFCD07787394
CAS Number: 3034-55-7
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR