5-Bromoquinoxaline 97%

Supplier: Ambeed
Warning

A201311-100MG A201311-250MG A201311-1G
103817-284EA 47 USD
103817-284 103847-922 76770-154 76770-156 77085-994
5-Bromoquinoxaline 97%
5-Bromoquinoxaline

Precaución: for Research Use Only

Formula: C₈H₅BrN₂
Boiling Pt: 300.2 °C (760 mm Hg)
Melting Pt: 66…67 °C
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD02181615
CAS Number: 76982-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR