5-Bromoquinoline-8-carbonitrile 98%

Supplier: Ambeed
Warning

A160751-100MG A160751-250MG A160751-1G A160751-25G A160751-5G
103805-416EA 32 USD
103805-416 103843-650 76812-720 76812-722 77300-496 77300-498 77300-500
5-Bromoquinoline-8-carbonitrile 98%
5-Bromoquinoline-8-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₅BrN₂
MW: 233.06 g/mol
Boiling Pt: 367.6±22.0 °C (760 mm Hg)
Flash Pt: 176.1 °C
Storage Temperature: Ambient
MDL Number: MFCD21340269
CAS Number: 507476-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR