5-Bromoquinoline-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A100982-250MG A100982-1G A100982-5G
103795-734EA 72 USD
103795-734 103821-426 76970-142 77282-630 77296-934
5-Bromoquinoline-2-carboxylic acid 97%
5-Bromoquinoline-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆BrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09881345
CAS Number: 1017412-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR