5-Bromoquinoline 97%

Supplier: Ambeed
Danger

A285357-1G A285357-5G A285357-10G A285357-25G A285357-100G
103813-672EA 9 USD
103813-672 103834-806 76837-340 76837-342 77262-700 77305-472 77616-218
5-Bromoquinoline 97%
5-Bromoquinoline

Precaución: for Research Use Only

Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 295.9 °C (760 mm Hg)
Melting Pt: 50…52 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00234481
CAS Number: 4964-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR