5-Bromo-8-quinolinol 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromoquinolin-8-ol

A309091-1G A309091-5G A309091-10G A309091-250MG
103814-586EA 45 USD
103814-586 103835-764 76838-530 76838-532 77032-574 77032-576
5-Bromo-8-quinolinol 95%
5-Bromo-8-quinolinol

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MW: 224.06 g/mol
Boiling Pt: 362.7 °C (760 mm Hg)
Melting Pt: 127 °C
Flash Pt: 173.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00457011
CAS Number: 1198-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR