8-Amino-5-bromoquinoline 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromoquinolin-8-amine

A132810-10G A132810-250MG A132810-1G A132810-5G A132810-100G
103801-786EA 23 USD
103801-786 77258-540 77258-542 77298-582 77298-584 77566-590
8-Amino-5-bromoquinoline 97%
8-Amino-5-bromoquinoline

Precaución: For research use only.

Formula: C₉H₇BrN₂
Boiling Pt: 353.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01571832
CAS Number: 53472-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR