5-Bromoquinolin-4-ol 95%

Supplier: Ambeed
Warning

A208160-1G A208160-5G
103817-860EA 83 USD
103817-860 103848-546 77086-850 77086-852
5-Bromoquinolin-4-ol 95%
5-Bromoquinolin-4-ol

Precaución: for Research Use Only

Formula: C₉H₆BrNO
Boiling Pt: 370.7±22.0 °C (760 mm Hg)
Flash Pt: 178 °C
Storage Temperature: Ambient
MDL Number: MFCD08063201
CAS Number: 723283-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR