5-Bromoquinolin-2(1H)-one 96%

Supplier: Ambeed
Warning

A380452-100MG A380452-250MG
103826-670EA 132 USD
103826-670 103837-704 76876-668 76876-670
5-Bromoquinolin-2(1H)-one 96%
5-Bromoquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MW: 224.054 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11108678
CAS Number: 99465-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR