5-Bromoquinazolin-4-ol 95%
Supplier: Ambeed
|
Warning
|
A103659-100MG
A103659-10G
A103659-250MG
A103659-25G
A103659-1G
103796-594EA
32
USD
103796-594
103822-302
76809-410
76809-412
76809-414
76809-416
77283-140
5-Bromoquinazolin-4-ol 95%
5-Bromoquinazolin-4-ol
Precaución: for Research Use Only
Formula:
C₈H₅BrN₂O Boiling Pt: 430.2 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD11559036 CAS Number: 147006-47-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...