5-Bromopyrimidine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A208571-250MG A208571-1G A208571-25G A208571-5G
103817-878EA 12 USD
103817-878 103848-564 76975-432 77086-886 77086-888 77086-890
5-Bromopyrimidine-2-carboxylic acid 97%
5-Bromopyrimidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₃BrN₂O₂
MW: 203.00 g/mol
Boiling Pt: 419.7 °C (760 mm Hg)
Flash Pt: 207.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00496793
CAS Number: 37131-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR