5-Bromopyrimidin-4(3H)-one sulfate(2:1) 95%
Supplier: Ambeed
|
Warning
|
A103631-100MG
A103631-1G
103796-586EA
24
USD
103796-586
103822-292
76780-662
77309-164
5-Bromopyrimidin-4(3H)-one sulfate(2:1) 95%
5-Bromopyrimidin-4(3H)-one sulfate(2:1)
Precaución: for Research Use Only
Formula:
C₈H₈Br₂N₄O₆S MW: 448.0500 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13195360 CAS Number: 97234-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...