5-Bromopyrimidin-2-ol 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-hydroxypyrimidine

A237960-10G A237960-25G A237960-5G
103850-970EA 12 USD
103850-970 76815-318 77304-092 77304-094
5-Bromopyrimidin-2-ol 98%
5-Bromopyrimidin-2-ol

Precaución: for Research Use Only

Formula: C₄H₃BrN₂O
MW: 174.9800 g/mol
Boiling Pt: 371.3 °C (760 mm Hg)
Melting Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD00233952
CAS Number: 214290-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR