2-Amino-5-bromopyrimidine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromopyrimidin-2-ylamine, 5-Bromopyrimidin-2-amine

A167698-25G A167698-5G A167698-100G A167698-10G A167698-1G A167698-250MG A167698-500G
103844-470EA 8 USD
103844-470 77081-502 77081-504 77273-588 77273-590 77273-592 77273-594 77273-596
2-Amino-5-bromopyrimidine 97%
2-Amino-5-bromopyrimidine

Precaución: for Research Use Only

Formula: C₄H₄BrN₃
MW: 174 g/mol
Melting Pt: 241…243 °C
Flash Pt: 159.876 °C
Storage Temperature: Ambient
MDL Number: MFCD00012341
CAS Number: 7752-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR