5-Bromopyridine-3-sulfonyl chloride 98%

Supplier: Ambeed
Danger

A509179-10G A509179-250MG
103828-364EA 54 USD
103828-364 103839-464 76774-550 76774-552
5-Bromopyridine-3-sulfonyl chloride 98%
5-Bromopyridine-3-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₅H₃BrClNO₂S
Boiling Pt: 329.1 °C (760 mm Hg)
Melting Pt: 60…63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08235263
CAS Number: 65001-21-0
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR