5-Bromo-2,3-pyridinediol 95%

Supplier: Ambeed
Warning

A152721-10G A152721-250MG A152721-1G A152721-5G
103804-426EA 25 USD
103804-426 103842-598 76768-202 76768-204 77299-976 77299-978
5-Bromo-2,3-pyridinediol 95%
5-Bromo-2,3-pyridinediol

Precaución: for Research Use Only

Formula: C₅H₄BrNO₂
Boiling Pt: 487.1 °C (760 mm Hg)
Melting Pt: 249 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08276314
CAS Number: 34206-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR