5-Bromopyridine-2,3-dicarboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A242297-100MG
A242297-10G
A242297-250MG
A242297-1G
A242297-5G
103811-460EA
15
USD
103811-460
103851-272
77000-456
77000-458
77000-460
77090-442
77090-444
5-Bromopyridine-2,3-dicarboxylic acid 97%
5-Bromopyridine-2,3-dicarboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₄BrNO₄ Boiling Pt: 434.7 °C (760 mm Hg) Melting Pt: 165 °C Flash Pt: 216.691 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07364745 CAS Number: 98555-51-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...