3-Amino-5-bromopyridine 98%

Supplier: Ambeed
Warning

A149313-100G A149313-1G A149313-10G A149313-500G
103804-026EA 5 USD
103804-026 103842-144 77286-074 77299-722 77567-116 77615-204
3-Amino-5-bromopyridine 98%
3-Amino-5-bromopyridine

Precaución: For research use only.

Formula: C₅H₅BrN₂
Melting Pt: 65…69 °C
Storage Temperature: Ambient
MDL Number: MFCD01646060
CAS Number: 13535-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR