2-Amino-5-bromopyridine 98%

Supplier: Ambeed
Warning

A216085-1000G A216085-25G A216085-100G A216085-10G A216085-500G A216085-1G
103809-510EA 49 USD
103809-510 103818-532 103849-244 77087-812 77087-814 77275-310 77275-312 77275-314 77303-424 77303-426
2-Amino-5-bromopyridine 98%
2-Amino-5-bromopyridine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂
Melting Pt: 133…138 °C
Flash Pt: 93.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00006323
CAS Number: 1072-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR