5-Bromopyridazin-4-amine 98%

Supplier: Ambeed
Warning

A146491-100MG A146491-250MG A146491-1G
103803-672EA 125 USD
103803-672 103841-764 76984-478 76984-480 77299-526
5-Bromopyridazin-4-amine 98%
5-Bromopyridazin-4-amine

Precaución: for Research Use Only

Formula: C₄H₄BrN₃
MW: 173.99 g/mol
Boiling Pt: 354.9±22.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD18909369
CAS Number: 55928-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR