5-Bromopyrazolo[1,5-a]pyrimidine 97%
Supplier: Ambeed
|
Warning
|
A153438-10G
A153438-250MG
A153438-1G
A153438-5G
103804-516EA
42
USD
103804-516
103842-688
76783-474
76783-476
77318-346
77318-348
5-Bromopyrazolo[1,5-a]pyrimidine 97%
5-Bromopyrazolo[1,5-a]pyrimidine
Precaución: for Research Use Only
Formula:
C₆H₄BrN₃ MW: 198.02 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11042744 CAS Number: 1159981-95-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...