5-Bromopyrazolo[1,5-a]pyridine 97%
Supplier: Ambeed
|
Warning
|
A106733-100MG
A106733-250MG
A106733-1G
A106733-5G
103797-064EA
25
USD
103797-064
103822-822
76869-466
76869-468
77309-674
77309-676
5-Bromopyrazolo[1,5-a]pyridine 97%
5-Bromopyrazolo[1,5-a]pyridine
Precaución: for Research Use Only
Formula:
C₇H₅BrN₂ MW: 197.032 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD13189494 CAS Number: 1060812-84-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...