5-Bromo-2-pyrazinecarboxylic acid 96%

Supplier: Ambeed
Warning

A331872-10G A331872-25G A331872-5G A331872-250MG
103825-590EA 10 USD
103825-590 103836-556 76788-174 76839-546 76839-548 77872-477
5-Bromo-2-pyrazinecarboxylic acid 96%
5-Bromo-2-pyrazinecarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₃BrN₂O₂
Boiling Pt: 341 °C (760 mm Hg)
Flash Pt: 160.03 °C
Storage Temperature: Freezer
MDL Number: MFCD09909672
CAS Number: 876161-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR