2-Bromo-5-cyanopyrazine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromopyrazine-2-carbonitrile

A205952-10G A205952-250MG A205952-1G A205952-25G A205952-5G
103817-672EA 22 USD
103817-672 103848-344 76785-498 76785-500 77086-576 77086-578 77086-580
2-Bromo-5-cyanopyrazine 97%
2-Bromo-5-cyanopyrazine

Precaución: for Research Use Only

Formula: C₅H₂BrN₃
MW: 184 g/mol
Melting Pt: 264.7 °C
Flash Pt: 113.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09909652
CAS Number: 221295-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR