2,3-Diamino-5-bromopyrazine 95%

Supplier: Ambeed
Warning

A137679-10G A137679-250MG A137679-1G A137679-25G
103802-484EA 9 USD
103802-484 76767-612 76767-614 77315-462 77315-464
2,3-Diamino-5-bromopyrazine 95%
2,3-Diamino-5-bromopyrazine

Precaución: for Research Use Only

Formula: C₄H₅BrN₄
Boiling Pt: 357.1 °C (760 mm Hg)
Melting Pt: 212…216 °C
Flash Pt: 169.8 °C
Storage Temperature: Ambient
MDL Number: MFCD06245575
CAS Number: 89123-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR