2-Bromo-5-hydroxypyrazine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromopyrazin-2-ol
A161412-100G
A161412-10G
A161412-1G
A161412-25G
A161412-5G
103805-484EA
10
USD
103805-484
103843-722
77096-178
77096-180
77319-698
77319-700
77319-702
2-Bromo-5-hydroxypyrazine 97%
2-Bromo-5-hydroxypyrazine
Precaución: for Research Use Only
Formula:
C₄H₃BrN₂O MW: 174.9800 g/mol Boiling Pt: 444 °C (760 mm Hg) Melting Pt: 115…119 °C Flash Pt: 222.324 °C Storage Temperature: Ambient |
MDL Number:
MFCD06245330 CAS Number: 374063-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...