5-Bromopyridine-2-carbonitrile 98%

Supplier: Ambeed
Danger

A149341-10G A149341-1G A149341-25G A149341-5G A149341-100G
103804-030EA 5 USD
103804-030 103842-148 77272-922 77299-724 77317-636 77317-638 77748-040
5-Bromopyridine-2-carbonitrile 98%
5-Bromopyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₃BrN₂
Melting Pt: 128…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00234144
CAS Number: 97483-77-7
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR