5-Bromo-2-formylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromopyridine-2-carbaldehyde, 5-Bromopicolinaldehyde, 5-Bromopyridine-2-carboxaldehyde

A214718-100G A214718-10G A214718-250MG A214718-500G A214718-25G A214718-5G A214718-1G
103818-430EA 6 USD
103818-430 103849-134 76999-724 76999-726 76999-728 76999-730 77087-660 77087-662 77303-374
5-Bromo-2-formylpyridine 98%
5-Bromo-2-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₄BrNO
Boiling Pt: 237.7 °C (760 mm Hg)
Melting Pt: 93 °C
Flash Pt: 97.562 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04112535
CAS Number: 31181-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR