5-Bromo-N-methylpyridin-2-amine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-(N-methylamino)pyridine, 5-Bromo-2-methylaminopyridine

A143588-100MG A143588-10G A143588-250MG A143588-1G A143588-25G A143588-5G
103803-302EA 43 USD
103803-302 76767-916 76767-918 76767-920 77316-584 77316-586 77316-588
5-Bromo-N-methylpyridin-2-amine 97%
5-Bromo-N-methylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MW: 187.0400 g/mol
Boiling Pt: 244.5 °C (760 mm Hg)
Flash Pt: 102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08272096
CAS Number: 84539-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR