5-Bromonicotinonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromopyridine-3-carbonitrile, 3-Bromopyridine-5-carbonitrile, 5-Bromo-3-cyanopyridine, 3-Bromo-5-cyanopyridine

A213280-1G A213280-100G A213280-25G A213280-5G A213280-10G
103818-284EA 9 USD
103818-284 103848-990 77087-492 77152-492 77303-278 77303-280 77685-568
5-Bromonicotinonitrile 98%
5-Bromonicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₃BrN₂
MW: 183.01 g/mol
Boiling Pt: 228.8 °C (760 mm Hg)
Melting Pt: 103…107 °C
Flash Pt: 92.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00174363
CAS Number: 35590-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR