3-Bromo-5-formylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromopyridine-5-carboxaldehyde, 5-Bromopyridine-3-carboxaldehyde, 5-Bromonicotinaldehyde, 5-Bromo-3-pyridinecarboxaldehyde

A178358-100G A178358-10G A178358-250MG A178358-500G A178358-1G A178358-25G A178358-5G
103807-324EA 8 USD
103807-324 103845-664 76998-482 76998-484 76998-486 76998-488 77083-082 77083-084 77083-086
3-Bromo-5-formylpyridine 98%
3-Bromo-5-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₄BrNO
MW: 186.01 g/mol
Melting Pt: 98 °C
Flash Pt: 105.7 °C
Storage Temperature: Ambient
MDL Number: MFCD03265758
CAS Number: 113118-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR