1-Amino-5-bromonaphthalene 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromonaphthalen-1-amine, 5-Bromo-1-naphthylamine
A216697-1G
A216697-25G
A216697-5G
A216697-100G
A216697-10G
A216697-500G
103818-576EA
10
USD
103818-576
103849-298
77087-890
77087-892
77087-894
77097-842
77097-844
77097-846
1-Amino-5-bromonaphthalene 97%
1-Amino-5-bromonaphthalene
Precaución: for Research Use Only
Formula:
C₁₀H₈BrN MW: 222.081 g/mol Boiling Pt: 358 °C (760 mm Hg) Melting Pt: 66…68 °C Flash Pt: 170.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD07369883 CAS Number: 4766-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...