5-Bromo-2-(dimethylamino)pyridine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-N,N-dimethylpyridin-2-amine
A201313-25G
A201313-250MG
A201313-10G
103817-286EA
8
USD
103817-286
103847-924
77085-996
77097-400
77685-509
5-Bromo-2-(dimethylamino)pyridine 97%
5-Bromo-2-(dimethylamino)pyridine
Precaución: for Research Use Only
Formula:
C₇H₉BrN₂ MW: 201.0600 g/mol Boiling Pt: 253.5 °C (760 mm Hg) Melting Pt: 41…44 °C Flash Pt: 107.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00099486 CAS Number: 26163-07-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...