5-Bromoisoquinoline 98%
Supplier: Ambeed
|
Warning
|
A195073-25G
A195073-5G
A195073-100G
A195073-500G
103816-720EA
7
USD
103816-720
103847-334
77085-282
77085-284
77260-622
77260-624
5-Bromoisoquinoline 98%
5-Bromoisoquinoline
Precaución: for Research Use Only
Formula:
C₉H₆BrN Boiling Pt: 312.3 °C (760 mm Hg) Melting Pt: 83…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD01646405 CAS Number: 34784-04-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...