5-Bromoisoquinolin-1(2H)-one 97%

Supplier: Ambeed
Warning

A447876-1G A447876-5G A447876-250MG A447876-25G A447876-10G
103827-768EA 30 USD
103827-768 103838-834 76842-640 76842-642 76979-468 76979-470 77673-104
5-Bromoisoquinolin-1(2H)-one 97%
5-Bromoisoquinolin-1(2H)-one

Precaución: For research use only

Formula: C₉H₆BrNO
Boiling Pt: 443.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11226915
CAS Number: 190777-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR