5-Bromoisophthalic acid 97%

Supplier: Ambeed
Warning

A116155-100G A116155-10G A116155-25G A116155-500G A116155-1G A116155-5G
103798-684EA 7 USD
103798-684 103824-548 77189-376 77189-378 77189-380 77189-382 77297-426 77297-428
5-Bromoisophthalic acid 97%
5-Bromoisophthalic acid

Precaución: for Research Use Only

Formula: C₈H₅BrO₄
MW: 245.03 g/mol
Boiling Pt: 452.1 °C (760 mm Hg)
Melting Pt: 276…287 °C
Flash Pt: 227.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00152019
CAS Number: 23351-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR