5-Bromoisoindoline hydrochloride 97%

Supplier: Ambeed
Warning

A196665-10G A196665-250MG A196665-1G A196665-25G A196665-5G
103816-860EA 29 USD
103816-860 103847-474 76975-088 76975-090 77085-440 77085-442 77085-444
5-Bromoisoindoline hydrochloride 97%
5-Bromoisoindoline hydrochloride

Precaución: for Research Use Only

Formula: C₈H₉BrClN
Storage Temperature: Ambient
MDL Number: MFCD10700135
CAS Number: 919346-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR