5-Bromo-1-isoindolinone 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2,3-dihydro-1H-isoindol-1-one, 5-Bromoisoindolin-1-one

A160909-100G A160909-10G A160909-1G A160909-25G A160909-5G
103805-432EA 15 USD
103805-432 103843-670 76783-830 76783-832 77319-632 77319-634 77319-636
5-Bromo-1-isoindolinone 98%
5-Bromo-1-isoindolinone

Precaución: for Research Use Only

Formula: C₈H₆BrNO
MW: 212.04 g/mol
Boiling Pt: 448.4 °C (760 mm Hg)
Melting Pt: 263…267 °C
Flash Pt: 224.965 °C
Storage Temperature: Ambient
MDL Number: MFCD09701292
CAS Number: 552330-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR