5-Bromoisatoic anhydride 95%

Supplier: Ambeed
Warning

A248635-25G A248635-5G A248635-100G A248635-10G A248635-1G
103811-832EA 9 USD
103811-832 103851-666 77091-016 77091-018 77193-608 77193-610 77304-452
5-Bromoisatoic anhydride 95%
5-Bromoisatoic anhydride

Precaución: for Research Use Only

Formula: C₈H₄BrNO₃
Boiling Pt: 526.9 °C (760 mm Hg)
Melting Pt: 280…285 °C
Flash Pt: 272.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00016921
CAS Number: 4692-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR