5-Bromoisatin 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromoindoline-2,3-dione, 5-Bromo-1H-indole-2,3-dione

A194051-25G A194051-5G A194051-1000G A194051-100G A194051-10G A194051-1G A194051-500G
103847-222EA 6 USD
103847-222 77085-134 77085-136 77274-552 77274-554 77274-556 77274-558 77274-560
5-Bromoisatin 98%
5-Bromoisatin

Precaución: for Research Use Only

Formula: C₈H₄BrNO₂
Melting Pt: 247…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00149345
CAS Number: 87-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR