5-Bromoindoline 98%

Supplier: Ambeed
Warning

A224137-10G A224137-1G A224137-25G A224137-5G
103819-086EA 17 USD
103819-086 103849-976 76691-890 77088-612 77088-614 77088-616
5-Bromoindoline 98%
5-Bromoindoline

Precaución: for Research Use Only

Formula: C₈H₈BrN
MW: 198.06 g/mol
Boiling Pt: 282.5 °C (760 mm Hg)
Melting Pt: 36…40 °C
Flash Pt: 110 °C
Storage Temperature: Freezer
MDL Number: MFCD00027410
CAS Number: 22190-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR