5-Bromooxindole 97%

Supplier: Ambeed
Warning

A262692-1G A262692-25G A262692-100G A262692-10G A262692-500G A262692-5G
103812-574EA 7 USD
103812-574 103852-470 77092-090 77092-092 77276-594 77276-596 77276-598 77304-856
5-Bromooxindole 97%
5-Bromooxindole

Precaución: for Research Use Only

Formula: C₈H₆BrNO
Boiling Pt: 362.5 °C (760 mm Hg)
Melting Pt: 220…224 °C
Flash Pt: 173.018 °C
Storage Temperature: Ambient
MDL Number: MFCD00456998
CAS Number: 20870-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR