5-Bromoimidazo[1,2-a]pyridine-2-carbaldehyde 95%

Supplier: Ambeed
Warning

A141353-100MG A141353-1G A141353-250MG
103802-980EA 46 USD
103802-980 76984-204 77316-132 77611-288
5-Bromoimidazo[1,2-a]pyridine-2-carbaldehyde 95%
5-Bromoimidazo[1,2-a]pyridine-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD11226918
CAS Number: 878197-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR