5-Bromo-2-furaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromofuran-2-carbaldehyde, 5-Bromofurfural

A146012-100G A146012-10G A146012-1G A146012-25G A146012-5G
103803-616EA 9 USD
103803-616 103841-702 77272-800 77272-802 77299-490 77317-008 77317-010
5-Bromo-2-furaldehyde 98%
5-Bromo-2-furaldehyde

Precaución: for Research Use Only

Formula: C₅H₃BrO₂
MW: 174.98 g/mol
Boiling Pt: 222.8 °C (760 mm Hg)
Melting Pt: 82…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00159501
CAS Number: 1899-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR