5-Bromochroman-4-one 97%
Supplier: Ambeed
|
Warning
|
A154179-100MG
A154179-250MG
A154179-1G
103804-612EA
400
USD
103804-612
103842-794
77150-560
77150-562
77318-492
5-Bromochroman-4-one 97%
5-Bromochroman-4-one
Precaución: for Research Use Only
Formula:
C₉H₇BrO₂ Boiling Pt: 339.4±42.0 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD09744050 CAS Number: 1199782-67-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...