5-Bromochroman 95%

Supplier: Ambeed
Warning

A139433-100MG A139433-1G
103802-740EA 364 USD
103802-740 76767-704 77315-834
5-Bromochroman 95%
5-Bromochroman

Precaución: for Research Use Only

Formula: C₉H₉BrO
MW: 213.07 g/mol
Boiling Pt: 274.6 °C (760 mm Hg)
Flash Pt: 108.7 °C
Storage Temperature: Ambient
MDL Number: MFCD08234478
CAS Number: 319478-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR