5-Bromobenzo[b]thiophene 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromobenzothiophene

A369346-100G A369346-10G A369346-500G A369346-5G A369346-1G A369346-25G
103826-450EA 11 USD
103826-450 103837-464 76773-456 76773-458 76773-460 76840-714 77306-804 77306-806
5-Bromobenzo[b]thiophene 97%
5-Bromobenzo[b]thiophene

Precaución: for Research Use Only

Formula: C₈H₅BrS
MW: 213.1 g/mol
Melting Pt: 47…48 °C
Storage Temperature: Ambient
MDL Number: MFCD03069318
CAS Number: 4923-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR