5-Bromobenzofuran-3(2H)-one 98%

Supplier: Ambeed
Warning

A111669-10G A111669-250MG A111669-25G A111669-1G A111669-5G
103797-882EA 103 USD
103797-882 103823-702 77160-778 77160-780 77160-782 77310-514 77310-516
5-Bromobenzofuran-3(2H)-one 98%
5-Bromobenzofuran-3(2H)-one

Precaución: for Research Use Only

Formula: C₈H₅BrO₂
Boiling Pt: 319.5 °C (760 mm Hg)
Flash Pt: 147.02 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06738724
CAS Number: 54450-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR