5-Bromobenzofuran 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromobenzo[b]furan

A212309-25G A212309-5G A212309-100G A212309-10G A212309-500G A212309-1G
103818-188EA 5 USD
103818-188 103848-886 77087-356 77087-358 77192-674 77192-676 77192-678 77303-240
5-Bromobenzofuran 97%
5-Bromobenzofuran

Precaución: for Research Use Only

Formula: C₈H₅BrO
MW: 197.04 g/mol
Boiling Pt: 233.8 °C (760 mm Hg)
Melting Pt: 8 °C
Flash Pt: 95.2 °C
Storage Temperature: Ambient
MDL Number: MFCD03407317
CAS Number: 23145-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR