5-Bromobenzo[d]oxazol-2-amine 98%

Supplier: Ambeed
Warning

A317210-10G A317210-1G A317210-25G A317210-5G
103825-128EA 14 USD
103825-128 103836-060 76772-866 77306-156 77306-158 77306-160
5-Bromobenzo[d]oxazol-2-amine 98%
5-Bromobenzo[d]oxazol-2-amine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂O
MW: 213.031 g/mol
Boiling Pt: 340.4 °C (760 mm Hg)
Flash Pt: 159.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01664213
CAS Number: 64037-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR